Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | CHF 46.00 |
Über diesen Artikel
CHF 46.00
material
blue
feature
fits beaker size 1000 to 4000 mL
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS8882D
General description
Features and Benefits
- Vinyl-coated lead ring will not mark or scratch benchtops and labware
- O-Shape Style
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description fits beaker size, 1000 to 4000 mL, blue | description fits beaker size, 250 to 1000 mL, blue | description fits beaker size, 125 to 500 mL, blue | description fits beaker size, 500 to 2000 mL, blue |
| feature fits beaker size 1000 to 4000 mL | feature fits beaker size 250 to 1000 mL | feature fits beaker size 125 to 500 mL | feature fits beaker size 500 to 2000 mL |
| material blue | material blue | material blue | material blue |
| manufacturer/tradename Heathrow Scientific HS8882D | manufacturer/tradename Heathrow Scientific HS8882B | manufacturer/tradename Heathrow Scientific HS8882A | manufacturer/tradename Heathrow Scientific HS8882C |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Vinyl-Coated Lead Ring ("O" shape)
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?