Merck
F1149
Sigma-Aldrich
Carestream® X-Omat LS film
Synonym(e):
autoradiography film, xray film
Größe auswählen
50 EA
CHF 337.00
CHF 337.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
50 EA
CHF 337.00
About This Item
UNSPSC-Code:
41105341
NACRES:
NB.22
CHF 337.00
Check Cart for Availability
Größe
8 in. (19.5 cm) × 10 in. (25.5 cm)
Allgemeine Beschreibung
Kodak Life Science Imaging film is equivalent to XRP. Coated with emulsion on both sides of a blue-tinted base, produces higher quality images than XAR film (improved definition with less background fog), however, the exposure time is approximately doubled. Sheets of film are interleaved with sheets of paper.
Rechtliche Hinweise
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Oliver Wicht et al.
Journal of virology, 88(14), 7952-7961 (2014-05-09)
Isolation of porcine epidemic diarrhea coronavirus (PEDV) from clinical material in cell culture requires supplementation of trypsin. This may relate to the confinement of PEDV natural infection to the protease-rich small intestine of pigs. Our study focused on the role
Questions
  1. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.