Fortfahren mit
B8157
Nalgene® Flaschen, Stil 2006
polypropylene copolymer, capacity 60 mL, translucent, with cap
Synonym(e):
nalgene bottle, nalgene bottles, Nalgene 2006-0002, laboratory container, receptical, vessel
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
12 ea
Warenkorb auf Verfügbarkeit prüfen
CHF 13.70
Über diesen Artikel
NACRES:
NB.17
UNSPSC Code:
41121800
CHF 13.70
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polypropylene copolymer bottle (PPCO), round translucent bottle, translucent , polypropylene copolymer
joint
Joints threaded neck (20)
description
Autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2006-0002
bottle diam. × H
37 mm × 85 mm
capacity
60 mL
General description
For lab applications requiring excellent chemical resistance and autoclaving, with and without contents, use Nalgene®arrow-mouth PPCO bottles with Closure. These polypropylene copolymer bottles are translucent, providing good clarity for viewing contents. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features and Benefits
Runde Enghalsflasche aus Polypropylen.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
B8032B8532B8282
description
polypropylene copolymer, capacity 60 mL, translucent, with cap
description
polypropylene copolymer, capacity 30 mL, translucent, with cap
description
polypropylene copolymer, capacity 500 mL, translucent, with cap
description
polypropylene copolymer, capacity 125 mL, translucent, with cap
feature
with cap
feature
with cap
feature
with cap
feature
with cap
manufacturer/tradename
Nalgene® 2006-0002
manufacturer/tradename
Nalgene® 2006-0001
manufacturer/tradename
Nalgene® 2006-0016
manufacturer/tradename
Nalgene® 2006-0004
capacity
60 mL
capacity
30 mL
capacity
500 mL
capacity
125 mL
material
polypropylene copolymer bottle (PPCO), translucent , round translucent bottle
material
polypropylene copolymer bottle (PPCO), polypropylene cap, translucent
material
round bottle, polypropylene copolymer , polypropylene copolymer bottle (PPCO), translucent
material
round bottle, polypropylene copolymer bottle (PPCO), translucent
joint
Joints threaded neck (20)
joint
Joints threaded neck (20)
joint
Joints threaded neck (28)
joint
Joints threaded neck (24)
bottle diam. × H
37 mm × 85 mm
bottle diam. × H
34 mm × 61 mm
bottle diam. × H
73 mm × 170 mm
bottle diam. × H
50 mm × 101 mm
Still not finding the right product?
Explore all of our products under Nalgene® Flaschen, Stil 2006
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)