
4,4′,4′′-Tris(N-(naphthalen-2-yl)-N-phenyl-amino)-triphenylamine

Synonym(s):
DPTPCz
Empirical Formula (Hill Notation):
C33H22N4
CAS Number:
Molecular Weight:
474.55
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing