Skip to Content
Merck
All Photos(2)

Documents

932736

Sigma-Aldrich

4,4′,4′′-Tris(N-(naphthalen-2-yl)-N-phenyl-amino)-triphenylamine

≥99% (HPLC)

Synonym(s):

DPTPCz, Host

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H22N4
CAS Number:
Molecular Weight:
474.55
UNSPSC Code:
12352116
NACRES:
NA.23

grade

sublimed grade

Quality Level

Assay

≥99% (HPLC)

transition temp

transition temp 86 °C (Tg)

solubility

2-methyltetrahydrofuran: soluble
ethyl acetate: soluble

λmax

305 nm±5 nm in ethyl acetate
353 nm±5 nm in ethyl acetate

fluorescence

λem 416 nm±10 nm in ethyl acetate

Orbital energy

HOMO 5.7 eV 
LUMO 2.8 eV 

Looking for similar products? Visit Product Comparison Guide

Application

4,4′,4′′-Tris(N-(naphthalen-2-yl)-N-phenyl-amino)-triphenylamine, also known as DPTPCz, is used as host/TADF material in organic electronics. It is solution-processable, and quite popular for electroluminescent device, and other optoelectronic device, such as displays (OLED).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small-Molecule Emitters with High Quantum Efficiency: Mechanisms, Structures, and Applications in OLED Devices.
Wei Q, et al.
Advanced Optical Materials, 6(20) (2018)
Xiao-Ke Liu et al.
Physical chemistry chemical physics : PCCP, 14(41), 14255-14261 (2012-09-01)
Recently, bipolar host materials have attracted considerable attention because they can achieve balanced charge injection/transport in phosphorescent organic light emitting diodes (PhOLEDs) and consequently obtain excellent device performance. In this work, two bipolar host materials, namely, 3-(4,6-diphenyl-1,3,5-triazin-2-yl)-9-phenyl-9H-carbazole (DPTPCz) and 3-(4,6-diphenoxy-1,3,5-triazin-2-yl)-9-phenyl-9H-carbazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service