Skip to Content
Merck
All Photos(1)

Key Documents

1612594

USP

Sevoflurane Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,1,1,3,3,3-Hexafluoro-2-propanol, HFP, Hexafluoroisopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2CHOH
CAS Number:
Molecular Weight:
168.04
Beilstein:
1841007
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sevoflurane

manufacturer/tradename

USP

refractive index

n20/D 1.275 (lit.)

bp

59 °C (lit.)

mp

−4 °C (lit.)

density

1.596 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(C(F)(F)F)C(F)(F)F

InChI

1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H

InChI key

BYEAHWXPCBROCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sevoflurane Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Sevoflurane

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

closed cup - boils before flash

Flash Point(C)

closed cup - boils before flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sevoflurane
United States Pharmacopeia, 38(5), 4036-4036 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service