Skip to Content
Merck
All Photos(1)

Key Documents

PHL89198

Gallic acid

phyproof® Reference Substance

Synonym(s):

3,4,5-Trihydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2CO2H
CAS Number:
Molecular Weight:
170.12
Beilstein:
2050274
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

mp

251 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

OC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C7H6O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,8-10H,(H,11,12)

InChI key

LNTHITQWFMADLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • Antioxidant Activity in plant species: This study highlights Gallic acid′s role in assessing antioxidant activities within various plant species, offering insights into its biochemical potential and applications in enhancing natural product research (Ghorbani et al., 2023).
  • Nutraceutical properties: Discusses Gallic acid′s significance in evaluating the nutraceutical properties of Aegle marmelos, emphasizing its potential in pharmaceutical and food industry applications (Sharma et al., 2022).
  • Phytopharmacological application: Focuses on the phytopharmacological aspects of Gallic acid, exploring its bioactive compounds and their therapeutic applications, particularly in traditional medicine (Ahmad et al., 2021).
  • Anti-inflammatory effects: Investigates the estrogenic and anti-inflammatory effects of Gallic acid-rich extracts, supporting its use in biomedical research focused on hormone-related and inflammatory conditions (Slighoua et al., 2021).
  • Gallic acid as a immunomodulatory agent: Examines Gallic acid′s effectiveness in modulating immune responses in vitro, highlighting its potential as an immunomodulatory agent (Lunic et al., 2020).

Biochem/physiol Actions

Gallic acid is a water soluble phenolic acid present in grapes and in the leaves of many plants. Gallic acid esters, such as tannins, catechin gallates and aliphatic gallates are potent antioxidants in vitro. However, gallic acid itself also appears to have antioxidant, anticarcinogenic and antiangiogenic activity in vitro.

Other Notes

This compound is commonly found in plants of the genus: eucalyptus humulus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service