Skip to Content
Merck
All Photos(1)

Key Documents

802354

TSKgel® HPLC Columns Hardware and Accessories

L × O.D. × I.D. 150 cm × 1/16 in. × 0.01 in., pkg of 1 ea

Synonym(s):

Tubing, PEEK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105108

product name

TSKgel® Hardware and Accessories, L × O.D. × I.D. 150 cm × 1/16 in. × 0.01 in., pkg of 1 ea

material

PEEK

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 02354

technique(s)

LPLC: suitable

L × O.D. × I.D.

150 cm × 1/16 in. × 0.01 in.

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service