This product is an MCE membrane, mixed cellulose esters, comprised of cellulose acetate and nitrate.
58188
Cellulose Filter Membrane
pore size 0.22 μm, diam. 47 mm, pkg of 100
About This Item
Recommended Products
material
cellulose
packaging
pkg of 100
diam.
47 mm
pore size
0.22 μm
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Is this regenerated cellulose?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service