Key Documents
56142
Omnifit® Small Bore Connector
Three-way valve connector
About This Item
UNSPSC Code:
12000000
Pricing and availability is not currently available.
description
PTFE cones included
technique(s)
LPLC: suitable
bore
0.8 mm
General description
Omnifit® Small Bore Connectors offer the unique advantage of producing a very low dead volume connection with tubing of various diameters, through an O-ring compression system, a cone over tubing system or a void-filling cone with a standard gripper.
Features and Benefits
PTFE cone included.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)