Key Documents
22970
Adustable Single Port Mini-Vap Evaporator/Concentrator
Mini-Vap size 1 1/2 in. × 8 1/2 in. , adjustable,
single unit
Synonym(s):
Mini-Vap 1mv ea
About This Item
UNSPSC Code:
41122001
Pricing and availability is not currently available.
Agency
suitable for EPA OTM-45
packaging
pkg of 1 ea
Mini-Vap size
1 1/2 in. × 8 1/2 in. , adjustable
General description
The Mini-Vap includes a needle valve for fine metering of air or nitrogen drying gas. Ideal for a wide variety of needs in evaporation or concentration.
The adjustable Mini-Vap (22970) is a versatile concentrator/evaporator that can be used with a single container. The unique design allows for any size container, from the smallest vial to a 250 mL beaker.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.