Skip to Content
Merck
All Photos(13)

Key Documents

ZRB1628

Sigma-Aldrich

Anti-S100-B Antibody, clone 8F20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100 calcium-binding protein B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 10.71 kDa
observed mol wt ~10 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 8F20 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects S100-B.

Immunogen

His-tagged full-length recombinant human S100-B protein.

Application

Quality Control Testing
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected S100-B in Rat Brain tissue lysate.Tested applicationsWestern Blotting Analysis: A 1:10,000 dilution from a representative lot detected S100-B in mouse spinal cord, human brain, and A375 lysates.Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected S100-B in human cerebellum tissue sections.Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected S100-B in A375 cells.Flow Cytometry Analysis: 1 μg from a representative lot detected S100-B in one million C6 cells.Affinity Binding Assay: A representative lot of this antibody bound S-100 B protein with a KD of 3.2 x 10-7 in an affinity binding assay. Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-S100-B, clone 8F20 ZooMAb, Cat. No. ZRB1628, is a recombinant Rabbit monoclonal antibody that specifically targets S100-B and is tested in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

Protein S100-B (UniProt: P04271; also known as S-100 protein beta chain, S-100 protein subunit beta, S100 calcium-binding protein B) is encoded by the S100B gene (Gene ID: 6285) in human. S100-B is a member of the EF-hand calcium-binding protein superfamily that is widely distributed and conserved in the central nervous system of vertebrates. It exists in both heterodimeric and homodimeric forms. Existence of dimer of either two alpha chains, or two beta chains, or one alpha and one beta chain has been reported and dimerization seems to be essential for its biological activities. S100-B has been suggested to play a part in a variety of cellular processes, primarily via binding to key synaptic proteins and inhibiting their phosphorylation. The extracellular form of S100-B, secreted by astrocytes, is physiologically involved in the development and maintenance of CNS homeostasis. S100-B effects are closely dependent on its cellular levels. At nanomolar concentrations it is shown to enhance survival of neurons during development and stimulate neurite outgrowth in cerebral cortex neurons. It also reduces the loss of mitochondrial function and cell death. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Donato, R. (2001). Int. J. Biochem. Cell Biol 33(7); 637-668; Rezaei, O., et al. (2017). Iran J. Neurol. 16(2); 83-89).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service