Skip to Content
Merck
All Photos(15)

Key Documents

ZMS1064

Sigma-Aldrich

Anti-Dynein Antibody, clone 74.1 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

74.1, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~68.56 kDa
observed mol wt ~78 kDa

species reactivity

rat, human, mouse

species reactivity (predicted by homology)

Xenopus, bovine, sheep, fish, Drosophila

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 74.1 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects cytoplasmic dynein.

Immunogen

Purified native bovine brain cytoplasmic dynein.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Dynein in HEK293 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Dynein in A431 and PC12 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Dynein in HeLa, HepG2, and NIH 3T3 cells.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected Dynein in one million HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Dynein, clone 74.1 ZooMAb®, Cat. No. ZMS1064, is a recombinant Mouse monoclonal antibody that specifically targets Dynein and is tested for use in Flow Cytometry, Immunocytochemistry, and Western Blotting.

Target description

Cytoplasmic dynein 1 intermediate chain 1 (UniProt: Q29RQ3; also known as Cytoplasmic dynein intermediate chain 1, Dynein intermediate chain 1, cytosolic, DH IC-1) is encoded by the DYNC1I1 gene (Gene ID: 613724) in bovine. Cytoplasmic dynein complexes are microtubules-associated retrograde transport motors. Cytoplasmic dynein 1 It is the more abundant motor in cells, while cytoplasmic dynein 2 takes part in intraflagellar transport. It is a multi-subunit complex of 1.5 MDa composed of a homodimer of heavy chains, two intermediate chains, light-intermediate chains, and light chains. The heavy chain homodimer acts as the dynein complex core and binds microtubules to enable ATP-dependent cytoplasmic dynein movement. The other dynein subunits associate as homodimers and play regulatory roles in maintaining the complex stability, modulating its activity, and mediating its interaction with accessory and cargo proteins. Cytoplasmic dynein 1 intermediate chain 1 acts as one of the several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are involved in linking dynein to cargos and to adapter proteins that regulate dynein function. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service