Merck
All Photos(4)
Key Documents
Z742615
PAGE-Optimizer Sample Preparation Column
10 columns
About This Item
UNSPSC Code:
41123100
NACRES:
NA.56
Pricing and availability is not currently available.
feature
10 columns
packaging
pkg of 10 ea
manufacturer/tradename
G-BioSciences 786-947
storage temp.
2-8°C
General description
PAGE-Optimizer is a unique, spin column for the preparation of samples for SDS-PAGE. The columns contain a proprietary separation matrix that removes contaminants such as salts, detergents, cellular agents and other common buffering agents. PAGE-Optimizer helps eliminate smeared lanes, distorted and/ or smiling bands and swollen lanes. PAGE-Optimizer has also been shown to reveal protein bands previously masked by the above interfering agents.
Include: 10 columns
Legal Information
PAGE-Optimizer is a trademark of Geno Technology, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)