Key Documents
Z710881
Dri-gauze filter liner for Tunair shake flask
For 2.5 L flask
Synonym(s):
Dri-gauze Filter, Filter Liner for Shake Flask, Tunair Shake Flask Filter
About This Item
UNSPSC Code:
41122104
NACRES:
NB.22
material
polypropylene
sterility
non-sterile
packaging
case of 5
manufacturer/tradename
IBI SS-3019
General description
The Dri-Gauze filter is 0.22 μm and constructed of nitrocellulose. Each filter can be used 10 – 12 times. The Dri-Gauze filters can be autoclaved in the cap. It consists of five dri-gauze filter cap linings to accommodate a 300 ml flask. The filter lining in the cap assembly can be replaced by pinching the flanges of the inner-closure shell until they snap loose. Then pull them apart and remove the used lining. The lining is replaced by sandwiching it between two parts of the cap and placing the cap back together. Ensure the flanges from the inner piece snap into the mated grooves in the outer piece, while reassembling. This makes the cap assembly stays intact during use.
Legal Information
DRI-GAUZE is a trademark of IBI Scientific
Tunair is a trademark of IBI Scientific
Visit the Document Library
Customers Also Viewed
Slide 1 of 1