Key Documents
Z670138
StainTray slide staining system
StainTray with clear lid
Synonym(s):
humidity tray, immunohistochemistry staining tray, slide staining tray
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
material
PETG lid
packaging
pack of 1 ea
manufacturer/tradename
Simport M920-1
parameter
-20-60 °C temp. range
L × H × W
38 cm × 4.5 cm × 24 cm
General description
  • Holds up to 20 slides
  • Drain plug for easy emptying
  • 4 rubber feet for greater stability
  • Stackable for save space
These robust hunidity chambers offer a more user friendly approach to immunohistochemistry staining. This tray is suitable for routine staining requiring a humidity chamber. Wells between rails holds water securely without splashing. The plastic rails are covered with a polymer strip for perfectly holding slides even if tray is held at an angle. Rails are raised not only to avoid water touching the slides but to make them more easily retrievable. The base will also excess stain solution dripping from the slides.
Choose from one of the most recent versions:
Certificates of Analysis (COA)