Key Documents
Z379271
Sigma-Aldrich
Deepwell 96 well plate
non-sterile
Synonym(s):
deepwell multiplate, storage plate
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polypropylene
round bottom clear wells (translucent)
size:
96 wells
sterility:
non-sterile
binding type:
non-treated surface
Pricing and availability is not currently available.
material
clear polypropylene
round bottom clear wells (translucent)
sterility
non-sterile
packaging
pack of 24 ea
manufacturer/tradename
Bel-Art F37860-0000
maximum volume
2 mL
size
96 wells
working volume
2 mL
binding type
non-treated surface
General description
These high-form polypropylene 96 well plates expand the flexibility of multichannel or automated pipetting systems. Use for non-binding protein assays, enzyme or receptor assays, or with cell harvesters. The 2 mL per well capacity makes it possible to do serial dilution procedures that are impossible with normal multiwell plates.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)