Key Documents
Z370762
Sigma-Aldrich
Scienceware® Holdfast graduated cylinders with handle
volume 1000 mL
About This Item
UNSPSC Code:
41121805
NACRES:
NB.43
Pricing and availability is not currently available.
material
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F28461-1000
volume measuring range
1000 mL
General description
Autoclavable polypropylene with double scale and stable octagon base. The handle provides a safe, solid grip for more controlled pouring from these large-sized cylinders.
Legal Information
Holdfast is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)