Key Documents
TMO3663-0250
Nalgene® graduated cylinders
PMP, volume 250 mL, case of 12 ea
Synonym(s):
PMP cylinder, graduated PMP cylinder
About This Item
UNSPSC Code:
41121805
NACRES:
NB.43
Pricing and availability is not currently available.
material
PMP
packaging
case of 12 ea
manufacturer/tradename
Nalgene 3663-0250
volume
250 mL
General description
Non-wetting, they deliver all they contain (calibrated TC/TD at 20°C). Meet Class B accuracy requirements of ASTM E1272 "Cylinder, Graduated, Laboratory, Glass" and all requirements of ISO Standard 6706 "Plastic Laboratory Ware-Graduated Measuring Cylinders". No meniscus to confuse readings. Blue, molded, PP base resists tipping. Molded-in graduations with additional major graduation lines on the back of cylinder for easier, more accurate fluid level readings. Generous pour spout. Can be autoclaved, but accuracy will be affected.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)