T6194
Poxygrid® 96 place test tube rack
Holds 96 x 15 mL tubes
Synonym(s):
tube holder
About This Item
Recommended Products
material
epoxy (coated steel)
feature
Holds 96 x 15 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18765-0001
L × W × H
24.2 cm (9¹/₂ in.) × 17 cm (6¹¹/₁₆ in.) × 6.4 cm (2¹/ in.)
General description
- Capable of quick freezing using liquid nitrogen
- Storage temperature: -80°C (freezer)
- Steam autoclavable: 121°C (250°F)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service