SYNF580810
Synthware™ Schlenk tube with high vaccum valve
capacity 10 mL, valve L 0-8 mm, O.D. × L 19 mm × 65 mm
Synonym(s):
Synthware F580810, Schlenk tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
clear glass
round bottom
sterility
non-sterile
feature
neck thread Internal
packaging
pack of 1 ea
manufacturer/tradename
Kemtech F580810
O.D. × L
19 mm × 65 mm
capacity
10 mL
valve L
0-8 mm
Looking for similar products? Visit Product Comparison Guide
General description
Schlenk Storage Tube, 10mL, ODxl 19X65mm includes Accessories: S600008T
Application
kemetch; synthware; tube; storage tube; schlenk tube
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service