Skip to Content
Merck
All Photos(1)

Key Documents

SRP4915

Sigma-Aldrich

Vaspin human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC)

Synonym(s):

OL-64, SERPINA12, Serpin A12 precursor, Vaspin, Visceral adipose tissue- derived serine protease inhibitor, Visceral adipose-specific serpin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (HPLC)
≥90% (SDS-PAGE)

form

lyophilized powder

mol wt

~47.0 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SPA12(145264)

General description

Vaspin (visceral adipose-specific SERPIN), a newly identified adipokine, is a member of serine protease inhibitor family. It is also known as serpin family A member 12 (SERPINA12). The gene encoding this protein is localized on human chromosome 14q32.13. Recombinant human Vaspin produced in E. coli is a single, non-glycosylated polypeptide chain containing 415 amino acids and having a molecular mass of 47kDa.

Biochem/physiol Actions

Vaspin (visceral adipose-specific SERPIN) is also a unique insulin sensitizing adipocytokine in obesity. A recent publication indicates that induction of human vaspin mRNA expression in adipose tissue is regulated in a fat depot-specific manner and could be associated with parameters of obesity, insulin resistance, and glucose metabolism. This protein may be useful as a biomarker with patients with acute myocardial infarction. It is a ligand of the 78kDa glucose-regulated protein/murine transmembrane protein (GRP78/MTJ-1) complex and has indirect effects on endoplasmic reticulum stress-induced metabolic disorders.

Physical form

1 mg/ml in 20 mM Tris pH-8, 0.2 mM PMSF and 10% glycerol.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feihong Yang et al.
Biochemical and biophysical research communications, 503(2), 501-507 (2018-05-08)
Visceral adipose tissue-derived serine protease inhibitor (vaspin), as a secretory adipokine, was reported to exert a protective role on insulin resistance. Recent studies showed that serum vaspin level was downregulated in patients with coronary artery disease. However, whether vaspin exerted
Vaspin as a Prognostic Marker in Patients with Acute Myocardial Infarction.
Zhang B
Heart, Lung & Circulation, 25(3), 257-264 (2016)
Vaspin: a novel adipokine, member of the family of serine protease inhibitors
Ekaterini Koiou
Aristotle University Medical Journal, 38.3, 7-18 (2011)
The role of vaspin in the development of metabolic and glucose tolerance disorders and atherosclerosis.
Dimova R and Tankova T
BioMed Research International, 823481-823481 (2015)
Jan Pippel et al.
Biological chemistry, 397(2), 111-123 (2015-11-04)
The adipokine vaspin (serpinA12) is mainly expressed in white adipose tissue and exhibits various beneficial effects on obesity-related processes. Kallikrein 7 is the only known target protease of vaspin and is inhibited by the classical serpin inhibitory mechanism involving a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service