Skip to Content
Merck
All Photos(1)

Key Documents

SRP0523

Sigma-Aldrich

COX1, FLAG® -His-Tags

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

Cox-1, Cyclooxygenase-1, PES-1, PGHS-1, PTGS1, Prostaglandin H2 Synthase 1, Prostaglandin-Endo-peroxide Synthase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

form

frozen liquid

mol wt

70.5 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... COX1(6775083)

General description

Human COX1, GenBank Accession No. NM_000962, a.a. 1-599 (end), with C-terminal FLAG-His-tag, MW=70.5 kDa, expressed in a baculovirus infected Sf9 cell expression system.

Physical form

Aqueous solution

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service