Skip to Content
Merck
All Photos(1)

Key Documents

SML4055

Sigma-Aldrich

UNC3230

≥98% (HPLC)

Synonym(s):

2-Anilino-5-(cyclohexanecarbonylamino)-1,3-thiazole-4-carboxamide, 5-(cyclohexanecarboxamido)-2-(phenylamino)thiazole-4-carboxamide, 5-[(Cyclohexylcarbonyl)amino]-2-(phenylamino)-4-thiazolecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O2S
CAS Number:
Molecular Weight:
344.43
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(N)C1=C(NC(C2CCCCC2)=O)SC(NC3=CC=CC=C3)=N1

InChI key

RZCNASHHHSKTGP-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of phosphatidylinositol 4-phosphate 5 kinase type 1C (PIP5K1C).



UNC3230 is a selective inhibitor of the lipid kinase phosphatidylinositol 4-phosphate 5 kinase type 1C (PIP5K1C) with an IC50 of 41 nM. UNC3230 showed selectivity for PIP5K1C and PIP4K2C (phosphatidylinositol 5-phosphate 4-kinase, type II, gamma) and did not inhibit PIP5K1A or any of the other lipid kinases that regulate phosphoinositide levels, including phosphatidylinositol 4,5-bisphosphate 3-kinases (PIK3s, also known as PI3Ks). UNC3230 lowered PIP2 levels in nociceptive dorsal root ganglia (DRG) neurons and had antinociceptive effects when injected intrathecally as well as peripherally into the hindpaw, indicating that either central or peripheral inhibition of PIP5K1C can reduce nociceptive responses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brittany D Wright et al.
Neuron, 82(4), 836-847 (2014-05-24)
Numerous pain-producing (pronociceptive) receptors signal via phosphatidylinositol 4,5-bisphosphate (PIP2) hydrolysis. However, it is currently unknown which lipid kinases generate PIP2 in nociceptive dorsal root ganglia (DRG) neurons and if these kinases regulate pronociceptive receptor signaling. Here, we found that phosphatidylinositol
Yukino Kato et al.
Experimental cell research, 405(1), 112654-112654 (2021-05-28)
Oligodendroglial cells (oligodendrocytes) differentiate to form the myelin that wraps neuronal axons in the central nervous system (CNS). This myelin sheath supports the propagation of saltatory conduction and protects axons from physical stresses. When oligodendrocytes do not normally differentiate to
Andrey A Parkhitko et al.
PLoS genetics, 17(2), e1009354-e1009354 (2021-02-17)
The RB1 tumor suppressor is recurrently mutated in a variety of cancers including retinoblastomas, small cell lung cancers, triple-negative breast cancers, prostate cancers, and osteosarcomas. Finding new synthetic lethal (SL) interactions with RB1 could lead to new approaches to treating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service