Skip to Content
Merck
All Photos(1)

Key Documents

SML3587

Sigma-Aldrich

SCH 900776

≥98% (HPLC)

Synonym(s):

6-Bromo-3-(1-methyl-1H-pyrazol-4-yl)-5-(3R)-3-piperidinyl-pyrazolo[1,5-a]pyrimidin-7-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18BrN7
CAS Number:
Molecular Weight:
376.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

NC1=C(Br)C([C@H]2CNCCC2)=NC3=C(C4=CN(C)N=C4)C=NN13

InChI

1S/C15H18BrN7/c1-22-8-10(6-19-22)11-7-20-23-14(17)12(16)13(21-15(11)23)9-3-2-4-18-5-9/h6-9,18H,2-5,17H2,1H3/t9-/m1/s1

InChI key

GMIZZEXBPRLVIV-SECBINFHSA-N

Biochem/physiol Actions

Highly potent and selective CHK1 inhibitor
SCH 900776 is a highly potent and selective CHK1 inhibitor that interacts synergistically with antimetabolites and other agents to induce cancer cell death.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting the replication checkpoint using SCH 900776, a potent and functionally selective CHK1 inhibitor identified via high content screening
Molecular Cancer Therapeutics, 10, 591-602 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service