Skip to Content
Merck
All Photos(1)

Key Documents

SML3438

Sigma-Aldrich

NDI-091143

≥90% (HPLC)

Synonym(s):

3-Chloro-5-[[(4,6-difluoro[1,1′-biphenyl]-3-yl)amino]sulfonyl]-4-hydroxy-benzoic acid, methyl ester, Methyl 3-chloro-5-[(2,4-difluoro-5-phenylphenyl) sulfamoyl]-4-hydroxybenzoate, NDI 091143, NDI091143

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14ClF2NO5S
CAS Number:
Molecular Weight:
453.84
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

Fc1c(cc(c(c1)F)c3ccccc3)N[S](=O)(=O)c2c(c(cc(c2)C(=O)OC)Cl)O

InChI

1S/C20H14ClF2NO5S/c1-29-20(26)12-7-14(21)19(25)18(8-12)30(27,28)24-17-9-13(15(22)10-16(17)23)11-5-3-2-4-6-11/h2-10,24-25H,1H3

InChI key

YSTSHUWHIDBZAK-UHFFFAOYSA-N

Biochem/physiol Actions

Highly potent, allosteric ATP-citrate lyase inhibitor.
NDI-091143 is a highly potent, allosteric ATP-citrate lyase inhibitor (human ACLY IC50 = 4.8 nM by coupled enzyme assay with 85 μM citrate; IC50 = 2.1 nM by ADP-Glo assay) that indirectly disrupts citrate binding (Ki = 7 nM) by targeting an adjacent hydrophobic site (kon = 1.02 x 10^6/M/s, koff = 0.0019/s).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service