Skip to Content
Merck
All Photos(1)

Key Documents

SML2954

Sigma-Aldrich

MGR2

≥98% (HPLC)

Synonym(s):

(Naphthalen-1-yloxy)methyl pivalate, 2,2-Dimethylpropanoic acid (1-naphthalenyloxy)methyl ester, MGR1 inactive control, MGR1 negative control

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O3
CAS Number:
Molecular Weight:
258.31
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to light brown

storage temp.

2-8°C

SMILES string

O(COC(=O)C(C)(C)C)c1c2c(ccc1)cccc2

InChI key

GLZRWJPKAZKJFB-UHFFFAOYSA-N

Biochem/physiol Actions

MGR2 is an inactive analog of and negative control for the reactive oxygen species (ROS) generator MGR1. Unlike MGR1, MGR2 does not induce cellular ROS or cause cell death at high concentrations (>5 μM; RAW264.7, THP1, HeLa, MCF7, A549).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service