Skip to Content
Merck
All Photos(1)

Key Documents

SML2500

Sigma-Aldrich

Senicapoc

≥98% (HPLC)

Synonym(s):

4-fluoro-α-(4-fluorophenyl)-α-phenyl-Benzeneacetamide, ICA 17043

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15F2NO
CAS Number:
Molecular Weight:
323.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N)C(C1=CC=C(F)C=C1)(C2=CC=C(F)C=C2)C3=CC=CC=C3

InChI

1S/C20H15F2NO/c21-17-10-6-15(7-11-17)20(19(23)24,14-4-2-1-3-5-14)16-8-12-18(22)13-9-16/h1-13H,(H2,23,24)

InChI key

SCTZUZTYRMOMKT-UHFFFAOYSA-N

Biochem/physiol Actions

Senicapoc (ICA-17043) is an inhibitor of the calcium-activated K+ channel KCa3.1, also known as the Gardos channel, which plays a role in K+ loss and dehydration of sickled red blood cells. Senicapoc blocks calcium-induced rubidium flux in human RBCs with and IC50 of 11 nM and prevents RBC dehydration in a transgenic mouse sickle cell disease model. It has been investigated for a number of possible uses, which include antimalarial activity, idiopathic pulmonary fibrosis, glioblastoma, lung cancer, non-alcoholic liver disease, hereditary xerocytosis, suppression of microglial activation in stroke, and immune system disorders.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service