Skip to Content
Merck
All Photos(1)

Key Documents

SML2402

Sigma-Aldrich

WYC-209

≥98% (HPLC)

Synonym(s):

2-[2-(3,4-Dihydro-4,4-dimethyl-1-oxido-2H-1-benzothiopyran-6-yl)ethynyl-5-pyrimidinecarboxylic acid, ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N2O3S
CAS Number:
Molecular Weight:
368.45
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S1CCC(C)(C)C2=C1C=CC(C#CC3=NC=C(C(OCC)=O)C=N3)=C2

InChI key

LLMDAYVUHYVJSS-UHFFFAOYSA-N

Biochem/physiol Actions

Synthetic retinoid with antiproliferative activity
WYC-209 is a synthetic retinoid with antiproliferative activity and better inhibition of cancer cell growth and proliferation than Tazarotene or ATRA (all-trans-retinoic acid). WYC-209 was shown to inhibit proliferation of malignant murine melanoma tumor-repopulating cells (TRCs) with an IC50 value of 190 nM and also inhibited proliferation of TRCs of human melanoma, lung cancer, ovarian cancer, and breast cancer in culture.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junwei Chen et al.
Nature communications, 9(1), 1406-1406 (2018-04-13)
Developing novel drugs that can abrogate the growth and metastasis of malignant tumors is a major challenge for cancer researchers. Here we describe a novel synthetic retinoid, namely WYC-209, which inhibits proliferation of malignant murine melanoma tumor-repopulating cells (TRCs), known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service