Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700120

Sigma-Aldrich

Monoclonal Anti-CD20, azide free antibody produced in mouse

clone MEM-97, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MS4A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-97, monoclonal

form

buffered aqueous solution

species reactivity

bovine, human, pig

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MS4A1(931)

General description

Cluster of differentiation 20 (CD20), also referred to as membrane spanning 4-domains A1 (MS4A1), is expressed on the surface of human B lymphocytes and on a small subset of T cells. The 33-35kDa, non-glycosylated protein has four membrane-spanning domains with both the amino and carboxy termini of the protein localized to the cytoplasm and a short 43 residue-extracellular segment. It is part of the membrane-spanning 4A gene family. The gene encoding MS4A1 is localized on human chromosome 11q12-13.

Immunogen

Raji human Burkitt′s lymphoma cell line

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 10 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Cluster of differentiation 20 (CD20) participates in the development and cell cycle progression in B-cells. It has been found to interact with proteins like adapter protein p75/80, CD40 and major histocompatibility complex class II proteins (MHC II). It may participate in Ca2+ influx across plasma membranes, maintenance of Ca2+ concentration and activation of B-cells. CD20 activates Src leading to rapid phosphorylation of phospholipase C-γ1 (PLC-γ1) and PLC-γ2, in turn activating caspase-3 signaling of apoptosis. Overexpression of the gene has been associated with the development of primary cutaneous T-cell lymphomas (CTCL).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From the bench to the bedside: ways to improve rituximab efficacy.
Cartron G
Blood, 104, 2635-2642 (2004)
Primary Cutaneous T-cell Lymphoma with Aberrant Expression of CD20.
Frings VG
Acta Dermato-Venereologica, 97, 534-536 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service