Skip to Content
Merck
All Photos(4)

Documents

SAB2701988

Sigma-Aldrich

Anti-KLF5 antibody produced in rabbit

Synonym(s):

BTEB2, CKLF, IKLF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

51 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: 500-10000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KLF5(688)

Immunogen

Recombinant fragment corresponding to a region within amino acids 69 and 346 of KLF5 (Uniprot ID#Q13887)

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, IP: Assay-dependent dilution, WB: 1:500-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This gene encodes a member of the Kruppel-like factor subfamily of zinc finger proteins. Since the protein localizes to the nucleus and binds the epidermal growth factor response element, it is thought to be a transcription factor. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geon-Ho Lee et al.
International journal of molecular sciences, 23(18) (2022-09-24)
The epithelial-mesenchymal transition (EMT) is a differentiation process associated with fibrogenesis in diabetic nephropathy (DN). Lysophosphatidic acid (LPA) is a small, naturally occurring glycerophospholipid implicated in the pathogenesis of DN. In this study, we investigated the role of LPA/LPAR1 signaling
Alejandro D Hofmann et al.
Pediatric surgery international, 30(12), 1191-1197 (2014-10-18)
The high morbidity and mortality in congenital diaphragmatic hernia (CDH) is attributed to pulmonary hypoplasia and persistent pulmonary hypertension (PH). PH is characterized by increased pulmonary artery smooth muscle cell (SMC) proliferation, suppressed apoptosis as well as endothelial dysfunction. Krüppel-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service