Skip to Content
Merck
All Photos(1)

Documents

SAB2500073

Sigma-Aldrich

Anti-APBA1/MINT1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Amyloid-β, Anti-D9S411E, Anti-MINT1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APBA1(320)

General description

APBA1 (Amyloid β (A4) precursor protein binding, family A, member 1) is a neuronal adaptor protein belonging to the X11 family. It consists of a centrally located phosphotyrosine binding domain which inhibits the production of Aβ peptide, two C-terminal postsynaptic density-95, large discs, zona occludens 1 (PDZ) domains.

Immunogen

Peptide with sequence C-EASHPSQDGKRQYK from the internal region of the protein sequence according to NP_001154.2.

Application

Anti-APBA1/MINT1 antibody produced in goat is suitable for indirect ELISA and western blot analysis.

Biochem/physiol Actions

APBA1 (Amyloid β (A4) precursor protein binding, family A, member 1) stabilizes amyloid precursor protein (APP) by directly interacting with it. It also restricts production as well as accumulation of proteolytic APP fragment in the brain of Alzheimer′s disease patients. It also suppresses glycogen synthase kinase-3β promoter activity by coupling with a silencer binding protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ju-Hyun Lee et al.
The Journal of biological chemistry, 278(47), 47025-47029 (2003-09-13)
Increased production and deposition of the 40-42-amino acid beta-amyloid peptide (Abeta) is believed to be central to the pathogenesis of Alzheimer's disease. Abeta is derived from the amyloid precursor protein (APP), but the mechanisms that regulate APP processing to produce
Kwok-Fai Lau et al.
Neuroreport, 21(11), 761-766 (2010-06-10)
X11alpha is a neuronal adaptor protein that interacts with the amyloid precursor protein (APP) through a centrally located phosphotyrosine binding domain to inhibit the production of Abeta peptide that is deposited in Alzheimer's disease brains. X11alpha also contains two C-terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service