Skip to Content
Merck
All Photos(1)

Key Documents

S0752

Sigma-Aldrich

(±)-Synephrine

≥98%

Synonym(s):

1-(4-Hydroxyphenyl)-2-methylaminoethanol, 4-Hydroxy-α-(methylaminomethyl)benzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH(CH2NHCH3)OH
CAS Number:
Molecular Weight:
167.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

mp

187 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

OC1=CC=C(C(O)CNC)C=C1

InChI

1S/C9H13NO2/c1-10-6-9(12)7-2-4-8(11)5-3-7/h2-5,9-12H,6H2,1H3

InChI key

YRCWQPVGYLYSOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(±)-Synephrine has been used as an alkaloid to investigate its in vitro antiviral, antibacterial, antifungal and cytotoxicity activities.

Biochem/physiol Actions

Synephrine is naturally found in citrus plants belonging to the Rutaceae family. It is a sympathomimetic alkaloid. Synephrine has 3 isomeric forms, such as m-synephrine, p-synephrine and o-synephrine. It exhibits β-adrenergic properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shun-ichi Ishiuchi et al.
The journal of physical chemistry. A, 115(37), 10363-10369 (2011-08-09)
In our previous work, we found that synephrine has six conformers in the gas phase, while adrenaline, which is a catecholamine and has the same side chain as synephrine, has been reported to have only two conformers. To determine the
C M Brown et al.
British journal of pharmacology, 93(2), 417-429 (1988-02-01)
1. The activities of the (-)- and (+)-forms of m- and p-octopamine and m- and p-synephrine on alpha 1-adrenoceptors from rat aorta and anococcygeus and alpha 2-adrenoceptors from rabbit saphenous vein were compared with those of noradrenaline (NA). 2. The
Jie-Ping Fan et al.
Analytical and bioanalytical chemistry, 402(3), 1337-1346 (2011-11-17)
In this work, molecularly imprinted solid-phase extraction (MISPE) has been used to selectively enrich, purify, or remove synephrine from Aurantii Fructus Immaturus. To this end, a molecularly imprinted polymer (MIP) was prepared by self-assembly from the template synephrine, the functional
C Carpéné et al.
Naunyn-Schmiedeberg's archives of pharmacology, 359(4), 310-321 (1999-05-27)
Numerous synthetic agonists selectively stimulate beta3-adrenoceptors (ARs). The endogenous catecholamines, noradrenaline and adrenaline, however, stimulate all the beta-AR subtypes, and no selective physiological agonist for beta3-ARs has been described so far. The aim of this study was to investigate whether
Comprehensive Natural Products II: Chemistry and Biology (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service