Skip to Content
Merck
All Photos(1)

Key Documents

K4518

Sigma-Aldrich

PKCγ, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC57564, PKC-gamma, PKCC, PKCG, PRK CG, SCA14

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

435-589 nmol/min·mg

mol wt

~105 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKCG(5582)

General description

PKCγ (protein kinase C γ) is a s a serine/threonine kinase belonging to the protein kinase C (PKC) family. PKCγ is mainly present in the central nervous system and is strongly expressed in cerebellar Purkinje cells.

Biochem/physiol Actions

PKCγ (protein kinase C γ) is involved in cell proliferation, differentiation as well as migration. Mutations in PKCγ are associated with neurodegenerative disease, spinocerebellar ataxia type 14. Single nucleotide polymorphisms in PKCγ are also linked with high risk of osteosarcoma.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Shamloo et al.
Neuroreport, 10(5), 931-935 (1999-05-13)
Neurons can be preconditioned against ischemic damage by a brief sublethal period of ischemia, applied several days before the second insult. Here we report on changes in the distribution and the levels of protein kinase Cgamma (PKCgamma) in nonconditioned and
Ying Zhang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(12), 12671-12677 (2014-09-26)
The objective of this study was to explore the relationship between single-nucleotide polymorphisms (SNPs) of the protein kinase C gamma (PRKCG) gene and osteosarcoma susceptibility in Chinese Han population. A total of 610 cases of osteosarcoma patients and 610 healthy
Hideyuki Takahashi et al.
Human molecular genetics, 24(2), 525-539 (2014-09-14)
Amyloid assemblies are associated with a wide range of human disorders, including Alzheimer's and Parkinson's diseases. Here, we identify protein kinase C (PKC) γ, a serine/threonine kinase mutated in the neurodegenerative disease spinocerebellar ataxia type 14 (SCA14), as a novel
Yi Liu et al.
The Biochemical journal, 478(10), 1999-2017 (2021-05-08)
Human hepatic tryptophan 2,3-dioxygenase (hTDO) is a homotetrameric hemoprotein. It is one of the most rapidly degraded liver proteins with a half-life (t1/2) of ∼2.3 h, relative to an average t1/2 of ∼2-3 days for total liver protein. The molecular mechanism
J Rennecke et al.
European journal of biochemistry, 242(2), 428-432 (1996-12-01)
13 murine tissues and 12 cell lines were tested for the expression of the novel protein kinase C (PKC) isoenzyme mu. Using two different PKC mu antibodies (sc-639 and P26720), PKC mu was detected in all tissues and cells and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service