Skip to Content
Merck
All Photos(2)

Key Documents

HT109

Sigma-Aldrich

Weigert′s Iron Hematoxylin Solution, Part B

Part B

Synonym(s):

Weigert’s iron hematoxylin solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116121
NACRES:
NA.47
Pricing and availability is not currently available.

Quality Level

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

To be used in conjunction with HT107. Recommended in lengthy or acidic stain procedures. Resists decolorization better than aluminum based hematoxylin formulations.

Other Notes

Ferric chloride in dilute HCl

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service