GE17549854
HiTrap® MabSelect™ PrismA
Cytiva, 17549854, pack of 5 × 5 mL
Synonym(s):
HiTrap MabSelect, MabSelect PrismA
About This Item
Recommended Products
packaging
pack of 5 × 5 mL
manufacturer/tradename
17549854
Cytiva
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The enhanced alkaline stability of MabSelect PrismA also provides opportunities for more robust mAb purification using periodic counter-current chromatography. The possibility for cleaning using 1 M NaOH increases bioburden control for continuous processes, which can be exposed to days or weeks of feed harvest.
Features and Benefits
- Enhanced binding capacity compared with other Protein A resins
- Excellent alkaline stability enables efficient cleaning and sanitization using 0.5–1.0 M NaOH
- Covered by a comprehensive security of supply program, including dual sources of the agarose base matrix and protein A ligand.
- The PrismA protein A ligand is also available in HiTrap® Fibro and HiScreen™ Fibro PrismA chromatography units for ultrafast purification of mAbs.
- Easily measure protein A leakage using the PrismA ELISA kit specifically designed for use with MabSelect™ PrismA resins.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
55.4 °F
Flash Point(C)
13 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service