Key Documents
EP5427750002
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-16-17, aerosol-tight, to hold, 16 x 5.0 mL Eppendorf tubes, with lid and wind shield
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Pricing and availability is not currently available.
feature
to hold 16 x 5.0 mL Eppendorf tubes
aerosol-tight
fixed angle rotor
with lid and wind shield
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427750002 (Global)
parameter
14,200 rpm
21,190 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)