EP5420000369
Eppendorf® Centrifuge 5420
without rotor, AC/DC input (GB plug)
Synonym(s):
Centrifuge
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
without rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5420000369
parameter
15,060 rpm speed
Looking for similar products? Visit Product Comparison Guide
General description
Centrifuge 5420 G, non- refrigerated, Keypad, without Rotor, 230V/50-60Hz, GB/HK
The Centrifuge 5420 was designed with your molecular biology routine applications in mind. The spectrum of methods includes DNA/RNA purification, DNA extraction from agarose gels, and PCR reaction set-up. It has a particularly small footprint to save space on the lab bench and an easy-to-clean keypad control panel. You can spin up to 24 tubes from 1.5 to 2.0 mL. Optional adapters extend the volume range to 0.4 and 0.5 mL tubes as well as 0.6 mL BD Microtainer. A rotor for 32 PCR tubes (0.2 mL) or four PCR strips is available as well.
Application
Centrifugation
Features and Benefits
- Standard rotor for up to 24 x 1.5 and 2.0 mL tubes
- Rotor option for up to 32 x 0.2 mL PCR tubes or 4 x 8-place PCR strips
- Adapters for 0.2, 0.4 and 0.5 mL tubes as well as 0.6 mL BD Microtainer
- Soft-touch one-finger lid closure for ergonomic lid locking
- Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
- High quality anodized aluminum rotors and rotor lids
- One push short-spin key for quick centrifugation - no need to hold key all the while
- Easy to clean keypad control panel
- Particularly small footprint to save precious space on the lab bench: 24 × 35 × 22 cm / 9,4 × 13,8 × 8,7 in (W × D × H)
- Max. speed: 21,300 × g (15,060 rpm)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service