Skip to Content
Merck
All Photos(1)

Key Documents

EP3881000023

Eppendorf® PCR cooler, 0.2 mL

capacity 0.2 mL, pink, pkg of 1 ea

Synonym(s):

Adjustable volume pipette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025
NACRES:
NC.21
Pricing and availability is not currently available.

material

pink

packaging

pkg of 1 ea

manufacturer/tradename

Eppendorf® 3881000023

capacity

0.2 mL

Looking for similar products? Visit Product Comparison Guide

General description

Get rid of the hassle of using ice buckets on your workstation and upgrade to our Eppendorf® PCR cooler, which is clean and easy to use. Ideal for protecting your samples during PCR setup.

Application

Eppendorf® PCR cooler, 0.2 mL is suitable for cooling of samples during the sample preparation step of polymerase chain reaction (PCR) method.

Features and Benefits

  • Eppendorf® PCR cooler, of capacity 0.2 mL is an accessory for liquid handling
  • It is a handling system for sample set-up, protection, transport, and storage of sensitive samples − keep your samples safe
  • It has a clear temperature indicator: the color of the PCR cooler changes when the temperature exceeds 7 °C.
  • Dry incubation technology reduces the contamination risk of samples
  • It has flexible vessel usage as it can accommodate PCR vessels as tubes, strips, or plates
  • The cooler can keep an entire 96-well PCR plate cold for more than an hour at 0 °C (with two-hour precooling at -20 °C) for safe samples

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service