EP3123000292
Eppendorf® Research plus G, Basic
single-channel, variable, volume 2-20 μL, light gray
About This Item
Recommended Products
feature
single-channel
variable
packaging
pack of 1 ea
pkg of 1 ea
manufacturer/tradename
Eppendorf® 3123000292
volume
2-20 μL
color
light gray
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Forward pipetting
- Reverse pipetting
- Removal of excess material
- Sample mixing
- Phase extraction
- Filling of plates, gels and reaction vessels
Features and Benefits
- Mechanical air cushion pipette with three-button operation as standard (one control for volume selection, one for pipetting and one for tip ejection)
- Ideal for accurate, easy and ergonomic pipetting of aqueous solutions by hand
- Convince yourself of the difference in weight and effort required when pipetting: Developed according to the principles of the Eppendorf PhysioCare Concept
- Benefit from one of the pipettes with the lowest operating and tip ejection forces on the pipette market to reduce hand and arm strain
- Minimized attachment forces thanks to a spring-loaded tip cone (available for all pipettes up to 1 mL)
- Adjust your pipette in seconds to improve accuracy when pipetting difficult liquids such as ethanol or when pipetting at high altitudes. Then return to factory adjustment without calibration
- Depending on your needs and application, autoclave the entire pipette or just the base to ensure proper decontamination
- High-quality material ensures high UV and chemical resistance; special single-channel bases available for aggressive chemicals
- Single-channel pipettes with ACT label and epT.IPS pipette tips help you make an informed choice when it comes to sustainability
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service