Skip to Content
Merck
All Photos(1)

Key Documents

EP0030108035

Eppendorf® DNA LoBind tubes

capacity 0.5 mL, PCR clean, withstand, 30,000 x g, pack of 250 ea

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11
Pricing and availability is not currently available.

material

cap (plug seal)
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube

sterility

non-sterile

feature

graduations
neck thread Push fit

packaging

pack of 250 ea

manufacturer/tradename

Eppendorf® 0030108035

technique(s)

PCR: suitable

capacity

0.5 mL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Maximize nucleic acid recovery. Eppendorf LoBind Tubes signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules. The ideal solution for sample preparation and long-term storage of your precious nucleic acids

Features and Benefits

  • Eppendorf® LoBind material ensures optimized sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • Available in tube formats for easy-up scaling.
  • Precise lid sealing for minimal evaporation rates in tube format.
  • Rated up to 30,000 × g centrifugation speed for molecular biology applications.

Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service