EP0030073819
Eppendorf® epT.I.P.S.® G Reloads, PCR clean
volume (2-200 μL), L (53 mm), yellow, pack of 960 ea (10 trays × 96 tips, stacked))
Synonym(s):
Reloadable pipette tips
About This Item
Recommended Products
material
yellow (Tray)
sterility
non-sterile
packaging
pack of 960 ea (10 trays × 96 tips, stacked))
manufacturer/tradename
Eppendorf® 0030073819
technique(s)
PCR: suitable
L
(53 mm)
maximum volume
200 μL
volume
(2-200 μL)
color
yellow
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Cell culture (media)
- Genomics: PCR, RT-PCR, qPCR, and all other types of PCR
- Enzymatic reactions (restriction decomposition, ligation)
- Isolation and purification of nucleic acid
- Gel electrophoresis (e.g., prefabricated DNA ladders) typical detergents: SDS, Triton™,
- X-100, Brij® 35, Tween® 20, CHAPS
- Proteomics (all types of protein examinations)
- Protein isolation and purification
- NGS library preparation
Features and Benefits
- Eppendorf® epT.I.P.S.® designed with the ultra-homogenous surface to provide maximized reproducibility
- Ultra hydrophobic surface of tips ensures minimal sample loss
- Minimizes foam formation during the pipetting process
- Highly resistant to chemicals
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service