Skip to Content
Merck
All Photos(1)

Key Documents

EHU077521

Sigma-Aldrich

MISSION® esiRNA

targeting human UBE2C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTCATGATGTCTGGCGATAAAGGGATTTCTGCCTTCCCTGAATCAGACAACCTTTTCAAATGGGTAGGGACCATCCATGGAGCAGCTGGAACAGTATATGAAGACCTGAGGTATAAGCTCTCGCTAGAGTTCCCCAGTGGCTACCCTTACAATGCGCCCACAGTGAAGTTCCTCACGCCCTGCTATCACCCCAACGTGGACACCCAGGGTAACATATGCCTGGACATCCTGAAGGAAAAGTGGTCTGCCCTGTATGATGTCAGGACCATTCTGCTCTCCATCCAGAGCCTTCTAGGAGAACCCAACATTGATAGTCCCTTGAACACACATGCTGCCGAGCTCTGGAAAAACCCCACAGCTTTTAAGAAGTACCTGCAAGAAACCTACTCAAAGCAGGTCACCAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei-Feng Liu et al.
Diagnostics (Basel, Switzerland), 10(9) (2020-09-10)
Ubiquitin-conjugating enzyme 2C (UBE2C) involves in numerous cellular processes and the tumor progression in many cancers. However, its role in oral squamous cell carcinoma (OSCC) is unclear. We aimed to investigate the role and clinical significance of UBE2C in OSCC.
Ying Wang et al.
Biochemical and biophysical research communications, 534, 597-603 (2020-11-24)
Ubiquitin Conjugating Enzyme E2 C (UBE2C) has a key oncogenic role in many human malignancies, including gastric cancer. However, it remains largely unknow at which level UBE2C expression is altered, as well as what are the downstream targets of UBE2C.
Liang Guo et al.
Cell cycle (Georgetown, Tex.), 16(18), 1705-1718 (2017-08-03)
Ubiquitin-conjugating enzyme E2C (UBE2C) is characterized as a crucial molecule in cancer cell growth that plays an essential role in the development of gliomas, but the detailed mechanisms have not been fully elucidated. In this study, we found that Forkhead
Xianxing Wang et al.
The FEBS journal, 286(24), 4889-4909 (2019-11-13)
Ubiquitin-conjugating enzyme 2C (UBE2C) is a core ubiquitin-conjugating enzyme in the ubiquitin-proteasome system that promotes cell cycle progression. Previous studies have indicated that UBE2C mediates tumorigenesis and progression in various cancers, but its role in pancreatic ductal adenocarcinoma (PDAC) remains
Rui Wang et al.
International journal of oncology, 50(4), 1116-1126 (2017-03-06)
The ubiquitin-conjugating enzyme 2C (UBE2C) is the key component in the ubiquitin proteasome system (UPS) by partnering with the anaphase‑promoting complex (APC/C). A high UBE2C protein expression level has been reported in various types of human tumors. However, little is known about the precise mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service