Skip to Content
Merck
All Photos(1)

Key Documents

CLS432058

Corning® CoolRack®

SV2, capacity 12 x 5 mL injectable ampules

Synonym(s):

Corning Coolcell accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122806
NACRES:
NB.12
Pricing and availability is not currently available.

material

foamed PE

feature

wells 12
Holds 12 x 5 mL tubes

packaging

pkg of 1

manufacturer/tradename

Corning 432058

technique(s)

cryopreservation: suitable

L × W × H

12.7 cm × 8.6 cm × 3.8 cm

size

27 mm , row spacing
30 mm , column spacing

well depth

24 mm

well diam.

16 mm

Looking for similar products? Visit Product Comparison Guide

General description

Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Certain CoolRack modules are SBS-compatible.

Legal Information

CoolRack is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service