CLS430917
Corning® microcentrifuge tubes with screw cap
2.0 mL microcentrifuge tube, polypropylene, w/ unattached rim seal screw cap, sterile, natural, 500/cs
Synonym(s):
centrifuge tubes, microcentrifuge tubes, microfuge tubes, screw cap microcentrifuge tubes
About This Item
Recommended Products
material
conical bottom tube
natural polypropylene tube
screw top cap
description
O-ring (no)
sterility
sterile
feature
disposable
self-standing conical bottom
packaging
pack of 50
case of 500
manufacturer/tradename
Corning 430917
parameter
20000 × g max. RCF
cap diam.
12.7 mm
capacity
2.0 mL
tube O.D.
10.7 mm
color
natural
fitting
cap (screw cap)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- 2 mL polypropylene microcentrifuge tubes feature screw caps that provide a tight secure seal
- Unattached rim seal cap
- Withstands a maximum RCF of 20,000xg
- Sterile
- Maximum RCF is 20,000
- No marking spot
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service