Key Documents
CLS3786
Corning® microcarriers
collagen coated, weight 10 g
Synonym(s):
lab equipment accessory
About This Item
UNSPSC Code:
41122100
NACRES:
NB.13
Pricing and availability is not currently available.
feature
collagen coated
packaging
pack of 1 ea
manufacturer/tradename
Corning 3786
weight
10 g
Related Categories
General description
Corning Microcarriers are designed to simplify your scale-up processes. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors.
Features:
  • USP Class VI polystyrene material provides a consistent platform
  • Level (SAL) of 10-6
  • Certified nonpyrogenic
These microcarriers are coated with Type I collagen to enhance cell attachment and expansion.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Articles
PLTMax® Human Platelet Lysate (hPL) is a superior serum-free and xeno-free media supplement alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) cultures.