CLS356500
Corning® BioCoat™ Collagen I 12 Well Clear Flat Bottom TC-Treated Multiwell Plate with Lid
well size 12 wells, flat bottom clear wells, lid
Synonym(s):
Cell Culture Plate, plastic coated cell culture plate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122102
NACRES:
NB.13
Recommended Products
material
flat bottom clear wells
feature
lid
packaging
pkg of 50 ea (10 packs of 50)
manufacturer/tradename
Corning 356500
well size
12 wells
binding type
Tissue Culture (TC)-treated surface
Application
Cell culture
Legal Information
BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service