CLS354414
Corning® BioCoat™ Poly-D-Lysine Multiwell Plate
flat bottom clear wells, lid
Synonym(s):
Cell Culture Plate, plastic coated cell culture plate
About This Item
Recommended Products
material
flat bottom clear wells
feature
lid
packaging
pkg of 5 ea (1 pack of 5)
manufacturer/tradename
Corning 354414
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | CLS354413 | CLS354509 | CLS356413 |
---|---|---|---|
lid | lid | lid | lid |
Tissue Culture (TC)-treated surface | Tissue Culture (TC)-treated surface | Tissue Culture (TC)-treated surface | Tissue Culture (TC)-treated surface |
flat bottom clear wells | flat bottom clear wells | flat bottom clear wells | flat bottom clear wells |
pkg of 5 ea (1 pack of 5) | pkg of 5 ea (1 pack of 5) | pkg of 5 ea (1 pack of 5) | pkg of 50 ea (10 packs of 50) |
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service