Skip to Content
Merck
All Photos(2)

Key Documents

CLS354259

Corning® Laminin/Entactin

from mouse(High Concentration), pack of 10.5 mg

Synonym(s):

extracellular matrix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116155
NACRES:
NA.7

biological source

mouse (High Concentration)

form

liquid

packaging

pack of 10.5 mg

manufacturer/tradename

Corning 354259

technique(s)

cell attachment: suitable
cell differentiation: suitable

Related Categories

General description

Laminin is a major component of basement membranes. It has numerous biological activities including promotion of cell adhesion, migration, chemotaxis, growth, and differentiation, including neurite outgrowth.
Molecular Weight
900 kD (unreduced)

Source
Engelbreth-Holm-Swarm (EHS) mouse tumor

Quality
Purity >/ 90% by SDS-PAGE
Tested for ability to initiate neurite outgrowth of NG-108 Mouse neuroblastoma cells
Found negative for the presence of bacteria, fungi, and mycoplasma

Preparation and Storage
Stable for at least three months at -70 °C. Avoid multiple freeze-thaws.

Handling
For a firm gel, a concentration of at least 3.5 mg/ml is recommended.

Formulation
High concentration laminin/entactin complex frozen in Dulbecco‘s PBS

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service