Skip to Content
Merck
All Photos(2)

Key Documents

CLS351177

Corning® Falcon® 96 Well Cell Culture Plate

round clear polystyrene, lid, sterile

Synonym(s):

Cell Culture Plate, Falcon 96-Well Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13

material

round clear polystyrene

sterility

sterile

feature

lid

manufacturer/tradename

Corning 351177

technique(s)

cell culture | mammalian: suitable

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service